Skip to Content
Merck
All Photos(1)

Key Documents

272345

Sigma-Aldrich

2,4,6-Triphenylpyrylium tetrafluoroborate

98%

Synonym(s):

2,4,6-Triphenylpyrylium fluoborate (6CI), 2,4,6-Triphenylpyrylium fluoroborate, 2,4,6-Triphenylpyrylium tetrafluoroborate (7CI), TPPT

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$133.00

$133.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$133.00

About This Item

Empirical Formula (Hill Notation):
C23H17BF4O
CAS Number:
Molecular Weight:
396.19
Beilstein:
3586533
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$133.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

mp

250-251 °C (lit.)

photocatalyst activation

462 nm

functional group

phenyl

SMILES string

F[B-](F)(F)F.c1ccc(cc1)-c2cc([o+]c(c2)-c3ccccc3)-c4ccccc4

InChI

1S/C23H17O.BF4/c1-4-10-18(11-5-1)21-16-22(19-12-6-2-7-13-19)24-23(17-21)20-14-8-3-9-15-20;2-1(3,4)5/h1-17H;/q+1;-1

InChI key

VQYPWMWEJGDSTF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4,6-Triphenylpyrylium tetrafluoroborate was used as a sensitizer to carry out the photooxidation of cathecol[1]. It was also used in the preparation of three N-alkylpyridinium photosensitizers by reacting with (1R,2S)-(-)-norephedrine, (S)-(+)-2-(aminomethyl)pyrrolidine and (R)-(-)-1-cyclohexylethylamine.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Experiences with replacement of the eyelid-levator].
K Hollmann
Fortschritte der Kiefer- und Gesichts-Chirurgie, 22, 98-99 (1977-01-01)
Mercedes Alvaro et al.
The Journal of organic chemistry, 67(15), 5184-5189 (2002-07-20)
Three N-alkylpyridinium photosensitizers having chiral alkyl groups have been prepared by reacting 2,4,6-triphenylpyrylium tetrafluoroborate ion with (1R,2S)-(-)-norephedrine, (S)-(+)-2-(aminomethyl)pyrrolidine, and (R)-(-)-1-cyclohexylethylamine. Laser flash photolysis allows detection of the corresponding triplet excited states that are quenched by hydrogen atom donors and electron
Remigiusz Bąchor et al.
International journal of molecular sciences, 21(9) (2020-05-07)
Detection of podocytes in urine might serve as a useful diagnostic tool in both primary and secondary glomerular diseases. The utility of podocyturia has been confirmed for both pre-eclampsia and glomerulonephritis. Here, we present a new and sensitive method for
Żaneta Rembisz et al.
Journal of chromatography. A, 1407, 184-192 (2015-07-15)
A high-performance liquid chromatography method based on pre-column derivatization with the pyrylium salts (4-[p-(N,N-dimethylamino)phenyl]-2,6-diphenylpyrylium perchlorate (LN1) and 2,4,6-triphenylpyrylium tetrafluoroborate (L1)) has been developed for the determination of sulfide. After the reaction of sulfide ions with LN1 or L1 aiming at

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service