Skip to Content
Merck
All Photos(1)

Key Documents

579963

Sigma-Aldrich

Diethylene glycol butyl ether

≥99%

Synonym(s):

Butyl CARBITOL, 2-(2-Butoxyethoxy)ethanol, BDG, Butyldiglycol

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$55.50
4 L
$67.60
18 L
$609.00

$55.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
$55.50
4 L
$67.60
18 L
$609.00

About This Item

Linear Formula:
CH3(CH2)3OCH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
162.23
Beilstein:
1739225
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
application(s):
hematology
bp:
231 °C (lit.)
vapor pressure:
30 mmHg ( 130 °C)

$55.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

5.6 (vs air)

Quality Level

vapor pressure

30 mmHg ( 130 °C)

Assay

≥99%

form

liquid

autoignition temp.

400 °F

expl. lim.

6.2 %

refractive index

n20/D 1.432 (lit.)

pH

7

bp

231 °C (lit.)

mp

−68 °C (lit.)

density

0.967 g/mL at 25 °C (lit.)

application(s)

hematology

functional group

ether
hydroxyl

SMILES string

CCCCOCCOCCO

InChI

1S/C8H18O3/c1-2-3-5-10-7-8-11-6-4-9/h9H,2-8H2,1H3

InChI key

OAYXUHPQHDHDDZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Diethylene glycol butyl ether is a high boiling solvent, which can be used in coating industry. It can also serve as a dispersant in organosols and can also be used in preparation of plasticizers.[1]

Application

  • Factors Affecting the Adsorption of Per- and Polyfluoroalkyl Substances (PFAS) by Colloidal Activated Carbon.: This study investigates the adsorption characteristics of PFAS by colloidal activated carbon, with a specific focus on the role of various solvents including diethylene glycol butyl ether. The research highlights the solvent′s effectiveness in enhancing adsorption efficiency (Hakimabadi et al., 2023).
  • First Stage of the Development of an Eco-Friendly Detergent Formulation for Efficient Removal of Carbonized Soil.: This research explores the formulation of eco-friendly detergents, demonstrating the use of diethylene glycol butyl ether as a key solvent for effective soil removal. The study underscores its potential in creating sustainable cleaning products (Fernandes et al., 2022).

Legal Information

Product of Dow Chemical
CARBITOL is a trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Diethylene Glycol Monobutyl Ether"
Gooch and Jan W
Encyclopedic Dictionary of Polymers, 216-216 (2011)
Thomas Göen et al.
Toxicology letters, 134(1-3), 295-300 (2002-08-23)
The aim of the study was to investigate the suitability of butoxyethoxyacetic acid (BEAA) as a biomarker of exposure to water-based cleaning agents containing diethylene glycol mono butyl ether (DEGBE). The study was performed in two printing plants where water-based
Yeomin Yoon et al.
Journal of membrane science, 261(1-2), 76-86 (2005-09-02)
Rejection characteristics of organic and inorganic compounds were examined for six reverse osmosis (RO) membranes and two nanofiltration (NF) membranes that are commercially available. A batch stirred-cell was employed to determine the membrane flux and the solute rejection for solutions
M Ema et al.
Drug and chemical toxicology, 11(2), 97-111 (1988-06-01)
The teratogenicity of diethylene glycol mono-n-butyl ether (DEGMBE) was studied in Wistar rats. The pregnant rats were fed a diet containing DEGMBE from day 0 through day 20 of pregnancy. The dietary concentrations of DEGMBE were 0, 0.04, 0.2 and
Christopher R Gibson et al.
Journal of the American Society for Mass Spectrometry, 14(11), 1247-1249 (2003-11-05)
Tandem liquid chromatography-mass spectrometry coupled to online radioactive material detection (LC/RAM/MS/MS) is a technique that is used routinely for in vivo and in vitro drug metabolism studies and allows for a simultaneous correlation between radiochemical peaks and mass spectral data.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service