Skip to Content
Merck
All Photos(1)

Documents

448281

Sigma-Aldrich

(3-Acrylamidopropyl)trimethylammonium chloride solution

75 wt. % in H2O

Synonym(s):

(Acrylamidopropyl)trimethylammonium chloride, Acryloylaminopropyltrimethylammonium chloride, Trimethyl(3-acrylamidopropyl)ammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCONH(CH2)3N(CH3)3Cl
CAS Number:
Molecular Weight:
206.71
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

contains

3,000 ppm monomethyl ether hydroquinone as stabilizer

concentration

75 wt. % in H2O

refractive index

n20/D 1.4848

density

1.11 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Cl-].C[N+](C)(C)CCCNC(=O)C=C

InChI

1S/C9H18N2O.ClH/c1-5-9(12)10-7-6-8-11(2,3)4;/h5H,1,6-8H2,2-4H3;1H

InChI key

OEIXGLMQZVLOQX-UHFFFAOYSA-N

General description

(3-Acrylamidopropyl)trimethylammonium chloride(APTAC) is a quaternary ammonium cationic monomer with high water solubility. It undergoes rapid cationic polymerization due to the presence of a permanent positive charge. It is utilized in the field of stimuli-responsive coatings, biosensors, drug delivery systems, medical devices, contact lenses, dental composites, wound healing, ion-exchange membranes for water treatment, and polymer 3Dscaffolds for tissue engineering.

Application

(3-Acrylamidopropyl)trimethylammonium chloride solution can be used:



  • As a cationic monomer to synthesize polyampholyte hydrogels (PAHs). These hydrogels show pH-responsive and ionic strength-responsive behavior and are suitable for application in smart coatings and biosensors.
  • As a monomer to prepare poly(APTAC) ion exchange membranes with high surface porosity, for the removal of Cu(II), Cr(VI), and As(V) ions from water.
  • As a precursor to fabricate cartilage mimetic polymer scaffolds that can be used in cartilage regeneration and replacement.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of cationic poly ((3-acrylamidopropyl) trimethylammonium chloride) by SARA ATRP in ecofriendly solvent mixtures
Mendoncca P, et al.
Polym. Chem., 5(19), 5829-5836 (2014)
Sarah Jurjevec et al.
Journal of colloid and interface science, 575, 480-488 (2020-05-16)
Macroporous polyampholyte hydrogels, simultaneously bearing both the anionic and cationic groups, demonstrate immense promise over the one-sign charged polyelectrolytes, owing to a unique phenomenon known as the ˝anti-polyelectrolyte˝ effect. Thus, they are extremely promising materials, since they remain solvated and
Novel biodegradable flocculanting agents based on pullulan
Ghimici L, et al.
Journal of Hazardous Materials, 181(1-3), 351-358 (2010)
Dorota Lachowicz et al.
Pharmaceutics, 11(12) (2019-11-27)
A cationic derivative of pullulan was obtained by grafting reaction and used together with dextran sulfate to form polysaccharide-based nanohydrogel cross-linked via electrostatic interactions between polyions. Due to the polycation-polyanion interactions nanohydrogel particles were formed instantly and spontaneously in water.
Emerging polymeric material strategies for cartilage repair
Connor J Demott and Melissa A Grunlan
Journal of materials chemistry. B, 10, 9578-9589 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service