Skip to Content
Merck
All Photos(1)

Key Documents

55707

Sigma-Aldrich

4-(4-Hydroxymethyl-3-methoxyphenoxy)butyric acid

≥98.0% (HPLC)

Synonym(s):

HMPB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16O5
CAS Number:
Molecular Weight:
240.25
Beilstein:
7876149
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98.0% (HPLC)

reaction suitability

reagent type: cross-linking reagent

impurities

≤5% water

mp

80-90 °C

SMILES string

COc1cc(OCCCC(O)=O)ccc1CO

InChI

1S/C12H16O5/c1-16-11-7-10(5-4-9(11)8-13)17-6-2-3-12(14)15/h4-5,7,13H,2-3,6,8H2,1H3,(H,14,15)

InChI key

HXOYWJCDYVODON-UHFFFAOYSA-N

Other Notes

Highly acid-labile linker for solid-phase peptide synthesis with FMOC protection

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debika Datta et al.
The journal of physical chemistry. B, 124(3), 577-588 (2019-12-28)
Peptide-based gels are emerging as an interesting class of biocompatible soft materials. 9-Fluorenylmethoxycarbonyl-protected amino acids and short peptides have gained considerable attention as promising gelators. Peptide amphiphiles, wherein an alkyl chain is appended to a polar peptidic moiety, are another
Diana P Slough et al.
The journal of physical chemistry. B, 122(14), 3908-3919 (2018-03-29)
Cyclic peptides are a promising class of molecules for unique applications. Unfortunately, cyclic peptide design is severely limited by the difficulty in predicting the conformations they will adopt in solution. In this work, we use explicit-solvent molecular dynamics simulations to
Xiaojun Cai et al.
ACS nano, 10(12), 11115-11126 (2016-12-28)
Prussian blue (PB) has been used as a photothermal conversion agent to generate heat to induce localized damage to tumor. However, its therapeutic efficiency is far from satisfactory. One of the major obstacles is that the maximum NIR absorption peak
B. Riniker et al.
Tetrahedron, 49, 9307-9307 (1993)
O. Goodings et al.
Journal of Combinatorial Chemistry, 1, 113-113 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service