Skip to Content
Merck
All Photos(1)

Key Documents

70921

Millipore

BugBuster® 10X Protein Extraction Reagent

BugBuster 10X is a concentrated formulation of the protein extraction reagent, using proprietary detergents employed in BugBuster without the addition of salts or buffer components.

Synonym(s):

BugBuster Reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106511
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

form

liquid

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

technique(s)

protein purification: suitable

suitability

suitable for column chromatography

application(s)

life science and biopharma

shipped in

ambient

storage temp.

10-30°C

General description

BugBuster 10X is a concentrated formulation of the proprietary detergents employed in BugBuster without the addition of salts or buffer components. Concentrated BugBuster provides a flexible alternative to the ready-to-use standard 1X BugBuster, allowing user-defined dilution and addition of buffer components. BugBuster 10X has all of the bioprocessing benefits of standard BugBuster plus the freedom to control pH, reagent concentration, and buffer additives necessary for maximum extraction and activity of your target protein.

Features and Benefits

Concentrated formulation: BugBuster 10X is a highly concentrated protein extraction reagent that offers a flexible alternative to the ready-to-use standard 1X BugBuster. This means that users can define their dilution and add buffer components to extract the maximum amount of target protein from the sample.

Proprietary detergents: The proprietary detergents used in BugBuster 10X provide all the benefits of the standard BugBuster reagent, such as efficient lysis of bacterial and yeast cells. The absence of salts and buffer components ensures that the extracted protein is of high purity and free from contaminants.

User-defined control: With BugBuster 10X, users have complete control over the pH, reagent concentration, and buffer additives necessary for optimal protein extraction and activity. This level of customization makes it an ideal solution for working on protein research with unique requirements.

Pain points addressed: BugBuster 10X addresses the pain points including but not limited to: low yields, inefficient extraction, and protein degradation. Its concentrated formulation and user-defined control offer a flexible and efficient solution to protein extraction, resulting in high-quality protein samples for downstream applications.

Warning

Toxicity: Standard Handling (A)

Legal Information

BUGBUSTER is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

ReadyShield® phosphatase and protease inhibitor cocktail FAQ for sample protection in a variety of cell types and tissue extracts, including mammalian, plant, and microbial samples. Our ReadyShield® Protease Inhibitor Cocktail is a non-freezing solution that contains inhibitors with a broad specificity for serine, cysteine, acid proteases and aminopeptidases.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service