Skip to Content
Merck
All Photos(1)

Key Documents

8.55061

Sigma-Aldrich

HMPB-MBHA resin

Novabiochem®

Synonym(s):

4-Hydroxymethyl-3-methoxyphenoxybutyric acid MBHA resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111023
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

alcohol

storage temp.

2-30°C

General description

This support consists of MBHA resin functionalized with Riniker′s hyperacid-labile 4-hydroxymethyl-3-methoxyphenoxybutyric acid linker [1].This hyperacid-sensitive resin is a useful tool for the Fmoc SPPS of protected peptide acid fragments. Attachment of carboxylic acids is normally achieved by DMAP catalyzed esterification with the appropriate symmetrical anhydride, or by using 2,6-dichlorobenzoylchloride/pyridine [2] or MSNT/Melm activation [3,4,5].Detachment of these functionalities can be effected by treatment with 1-5% TFA in DCM.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Linkage

Replaces: 01-64-0036

Analysis Note

Colour (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.40 - 1.00 mmol/g
Swelling Volume (in DMF): lot specific result
The polymer matrix is copoly ( styrene - 1% DVB ) 100- 200 mesh

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novabiochem® offers a wide range of linkers and derivatized resins for Fmoc solid-phase peptide synthesis with specialized protocols.

Protocols

Review various resins like Merrifield, trityl-based, and hydroxymethyl-functionalized for peptide immobilization for diverse applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service