Skip to Content
Merck
All Photos(1)

Key Documents

553190

Sigma-Aldrich

1-(2-Methylpropyl)-4-piperidone

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$319.00

$319.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$319.00

About This Item

Empirical Formula (Hill Notation):
C9H17NO
CAS Number:
Molecular Weight:
155.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$319.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.456 (lit.)

bp

220 °C (lit.)

density

0.914 g/mL at 25 °C (lit.)

functional group

ketone

storage temp.

2-8°C

SMILES string

CC(C)CN1CCC(=O)CC1

InChI

1S/C9H17NO/c1-8(2)7-10-5-3-9(11)4-6-10/h8H,3-7H2,1-2H3

InChI key

KMTASGXMTBUSSP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-(2-Methylpropyl)-4-piperidone is a piperidone derivative. It can be synthesized by employing enamine derived from isobutyraldehyde and piperidone as a starting reagent.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.9 °F - closed cup

Flash Point(C)

93.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry of phosphorous acid: new routes to phosphonic acids and phosphate esters.
Redmore D.
The Journal of Organic Chemistry, 43(5), 992-996 (1978)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service