Skip to Content
Merck
All Photos(1)

Key Documents

B29806

Sigma-Aldrich

1-Benzyl-4-piperidone

99%

Synonym(s):

1-Benzyl-4-oxopiperidine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$53.80
100 G
$104.00

$53.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$53.80
100 G
$104.00

About This Item

Empirical Formula (Hill Notation):
C12H15NO
CAS Number:
Molecular Weight:
189.25
Beilstein:
128556
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$53.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.541 (lit.)

bp

134 °C/7 mmHg (lit.)

density

1.021 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C1CCN(CC1)Cc2ccccc2

InChI

1S/C12H15NO/c14-12-6-8-13(9-7-12)10-11-4-2-1-3-5-11/h1-5H,6-10H2

InChI key

SJZKULRDWHPHGG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Benzyl-4-piperidone serves as a versatile heterocyclic building block in the synthesis of pharmaceutical compounds.[1][2]

Application

1-Benzyl-4-piperidone is generally used as a building block in the synthesis of various medicinal compounds. Applications include the synthesis of:
  • Spirocyclic furopyridines as haloperidol-sensitive σ receptor ligands.[3][4]
  • Multi-target-directed donepezil + propargylamine + 8-hydroxyquinoline (DPH) hybrids for the treatment of Alzheimer′s disease.[2]
  • Spiropiperidine iminohydantoin aspartyl protease inhibitors.[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F

Flash Point(C)

71 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel spiropiperidines as highly potent and subtype selective ?-receptor ligands. Part 1.
Maier CA and Wunsch B
Journal of Medicinal Chemistry, 45(2), 438-448 (2002)
Synthesis and ? receptor affinity of regioisomeric spirocyclic furopyridines.
Miyata K, et al.
European Journal of Medicinal Chemistry, 83(2), 709-716 (2014)
Donepezil+ propargylamine+ 8-hydroxyquinoline hybrids as new multifunctional metal-chelators, ChE and MAO inhibitors for the potential treatment of Alzheimer's disease.
Wang Li, et al.
European Journal of Medicinal Chemistry, 80(2), 543-561 (2014)
Di-tert-butylsilylene-directed α-selective synthesis of 4-methylumbelliferyl T-antigen.
Imamura A, et al.
Organic Letters, 7(20), 4415-4418 (2005)
Chunmei Gao et al.
Bioorganic & medicinal chemistry, 16(18), 8670-8675 (2008-08-21)
A novel series of 10-benzyl-9(10H)-acridinones and 1-benzyl-4-piperidones were synthesized and tested for their in vitro antitumor activities against CCRF-CEM cells. Assay-based antiproliferative activity study using CCRF-CEM cell lines revealed that the acridone group and the substitution pattern on the benzene

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service