Skip to Content
Merck
All Photos(1)

Key Documents

544264

Sigma-Aldrich

2,6-Difluorobenzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3SO2Cl
CAS Number:
Molecular Weight:
212.60
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.526 (lit.)

bp

210 °C (lit.)

density

1.568 g/mL at 25 °C (lit.)

SMILES string

Fc1cccc(F)c1S(Cl)(=O)=O

InChI

1S/C6H3ClF2O2S/c7-12(10,11)6-4(8)2-1-3-5(6)9/h1-3H

InChI key

QXWAUQMMMIMLTO-UHFFFAOYSA-N

General description

2,6-Difluorobenzenesulfonyl chloride can be prepared by reacting difluorophenyllithium with sulfuryl chloride.

Application

2,6-Difluorobenzenesulfonyl chloride may be used in the synthesis of 2,6-difluorobenzenesulfonamide and benzoxathiazocine 1,1-dioxides core scaffolds.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joanna K Loh et al.
Beilstein journal of organic chemistry, 8, 1293-1302 (2012-09-29)
The efficient synthesis of an 80-member library of unique benzoxathiazocine 1,1-dioxides by a microwave-assisted, intermolecular nucleophilic aromatic substitution (S(N)Ar) diversification pathway is reported. Eight benzofused sultam cores were generated by means of a sulfonylation/S(N)Ar/Mitsunobu reaction pairing protocol, and subsequently diversified
Insect growth regulators. Analogues of TH-6038 and TH-6040.
J E Oliver et al.
Journal of agricultural and food chemistry, 24(5), 1065-1068 (1976-09-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service