Skip to Content
Merck
All Photos(3)

Documents

545708

Sigma-Aldrich

2,6-Dichlorobenzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3SO2Cl
CAS Number:
Molecular Weight:
245.51
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

53-56 °C (lit.)

SMILES string

Clc1cccc(Cl)c1S(Cl)(=O)=O

InChI

1S/C6H3Cl3O2S/c7-4-2-1-3-5(8)6(4)12(9,10)11/h1-3H

InChI key

WGGKQIKICKLWGN-UHFFFAOYSA-N

General description

2,6-Dichlorobenzenesulfonyl chloride, also known as 2,6-dichlorophenylsulfonyl chloride, is an aryl sulfonyl chloride derivative.

Application

2,6-Dichlorobenzenesulfonyl chloride may be used as a starting material in the multi-step synthesis of sulfonamide-containing diarylsquaramide. It may also be used in the synthesis of cycloheptyl substituted 1,2,4-triazolopyridine (TZP) analogs.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haixia Wang et al.
Bioorganic & medicinal chemistry letters, 21(14), 4146-4149 (2011-06-22)
A series of pyridyl amide/sulfonamide inhibitors of 11β-HSD-1 were modified to incorporate a novel 1,2,4-triazolopyridine scaffold. Optimization of substituents at the 3 and 8 position of the TZP core, with a special focus on enhancing metabolic stability, resulted in the
Brent W McCleland et al.
Bioorganic & medicinal chemistry letters, 17(6), 1713-1717 (2007-01-24)
N,N'-diarylsquaramides were prepared and evaluated as antagonists of CXCR2. The compounds were found to be potent and selective antagonists of CXCR2. Significant differences in SAR was observed relative to the previously described N,N'-diarylurea series. As was the case in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service