Skip to Content
Merck
All Photos(1)

Documents

539775

Sigma-Aldrich

Ethane

99.99%

Synonym(s):

Ethane gas

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH3
CAS Number:
Molecular Weight:
30.07
Beilstein:
1730716
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

vapor density

1.05 (vs air)

vapor pressure

37.95 atm ( 21.1 °C)

Assay

99.99%

form

gas

autoignition temp.

881 °F

expl. lim.

13 %

bp

−88 °C (lit.)

mp

−172 °C (lit.)

density

0.362 g/mL at 20 °C (lit.)

SMILES string

CC

InChI

1S/C2H6/c1-2/h1-2H3

InChI key

OTMSDBZUPAUEDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • High-pressure oxidation of ethane: The paper discusses the oxidation properties of ethane under high pressure, providing insights crucial for developing combustion models and understanding ethane′s behavior in various industrial processes (H Hashemi, JG Jacobsen, CT Rasmussen, 2017).
  • Progress and prospects in catalytic ethane aromatization: This review highlights the advancements in converting ethane to more valuable aromatic hydrocarbons, showcasing the potential of ethane as a petrochemical feedstock (Y Xiang, H Wang, J Cheng, J Matsubu, 2018).

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-211.0 °F - closed cup

Flash Point(C)

-135 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reika Kanya et al.
The Journal of chemical physics, 136(20), 204309-204309 (2012-06-07)
Two-body Coulomb explosion processes of ethane (CH(3)CH(3)) and its isotopomers (CD(3)CD(3) and CH(3)CD(3)) induced by an intense laser field (800 nm, 1.0 × 10(14) W/cm(2)) with three different pulse durations (40 fs, 80 fs, and 120 fs) are investigated by
Paul O Wennberg et al.
Environmental science & technology, 46(17), 9282-9289 (2012-08-03)
We use historical and new atmospheric trace gas observations to refine the estimated source of methane (CH(4)) emitted into California's South Coast Air Basin (the larger Los Angeles metropolitan region). Referenced to the California Air Resources Board (CARB) CO emissions
Calvin C H Chan et al.
Environmental science & technology, 46(18), 10154-10160 (2012-08-21)
Compound specific isotope analysis (CSIA) has been applied to monitor bioremediation of groundwater contaminants and provide insight into mechanisms of transformation of chlorinated ethanes. To date there is little information on its applicability for chlorinated methanes. Moreover, published enrichment factors
Isobel J Simpson et al.
Nature, 488(7412), 490-494 (2012-08-24)
After methane, ethane is the most abundant hydrocarbon in the remote atmosphere. It is a precursor to tropospheric ozone and it influences the atmosphere's oxidative capacity through its reaction with the hydroxyl radical, ethane's primary atmospheric sink. Here we present
Margaret A Greene et al.
Organic letters, 14(16), 4293-4296 (2012-05-10)
Stereospecific nickel-catalyzed cross-coupling reactions of benzylic 2-methoxyethyl ethers are reported for the preparation of enantioenriched 1,1-diarylethanes. The 2-methoxyethyl ether serves as a traceless directing group that accelerates cross-coupling. Chelation of magnesium ions is proposed to activate the benzylic C-O bond

Protocols

Separation of Methane; Acetylene; Carbon monoxide; Water; Nitrogen; Carbon dioxide; Ethane; Ethylene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service