Skip to Content
Merck
All Photos(1)

Key Documents

740543

Sigma-Aldrich

1α,25-Dihydroxyvitamin D3 (6,19,19-d3) solution

50 μg/mL in ethanol, 97 atom % D, 96% (CP)

Synonym(s):

1α,25-Dihydroxycholecalciferol (6,19,19-d3) solution, Calcitriol (6,19,19-d3) solution, Deuterated 1-α25-Dihydroxyvitamin D3 solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$1,670.00

$1,670.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 ML
$1,670.00

About This Item

Empirical Formula (Hill Notation):
C27D3H41O3
CAS Number:
Molecular Weight:
419.65
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24

$1,670.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

97 atom % D

Quality Level

Assay

96% (CP)

form

solution

concentration

50 μg/mL in ethanol

technique(s)

mass spectrometry (MS): suitable

color

colorless

mp

-144.0 °C

application(s)

detection

mass shift

M+3

storage temp.

−20°C

SMILES string

[2H]/C([2H])=C1[C@@H](O)C[C@H](O)CC\1=C([2H])\C=C2/CCC[C@]3(C)[C@H](CC[C@@H]23)[C@H](C)CCCC(C)(C)O

InChI

1S/C27H44O3/c1-18(8-6-14-26(3,4)30)23-12-13-24-20(9-7-15-27(23,24)5)10-11-21-16-22(28)17-25(29)19(21)2/h10-11,18,22-25,28-30H,2,6-9,12-17H2,1,3-5H3/b20-10+,21-11-/t18-,22-,23-,24+,25+,27-/m1/s1/i2D2,11D

InChI key

GMRQFYUYWCNGIN-POLIPXRRSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

1α,25-Dihydroxyvitamin D3 [1,25(OH)2D3], also known as calcitriol, plays a role in calcium and phosphate metabolism in humans. It also regulates insulin secretion and induces cellular differentiation.[1][2]
1α,25-Dihydroxyvitamin D3 (6,19,19-d3) is a deuterated vitamin D3 wherein C-6 and C-19 protons are replaced by deuterium.

Application

1α,25-Dihydroxyvitamin D(6,19,19-d3) can be used as a stable isotope internal standard for accurate data interpretation in specific biological samples by mass spectrometry.

Packaging

1 mL in ampule

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianqiong Zheng et al.
Frontiers in pharmacology, 11, 97-97 (2020-03-19)
Metabolic syndrome is a disorder of energy use and storage, which is characterized by central obesity, dyslipidemia, and raised blood pressure and blood sugar levels. Maternal 25-hydroxyvitamin D deficiency is known to cause metabolic changes, chronic disease, and increased adiposity
Vitamin D3: a helpful immuno-modulator
Di R, et al.
Immunology, 134(2), 123-139 (2011)
The emerging role of vitamin D3 in women′s health
Khadilkar, SS
The Journal of Obstetrics and Gynecology of India, 63(2), 147?150-147?150 (2013)
Xinzhu Liu et al.
Burns & trauma, 8, tkaa042-tkaa042 (2021-01-28)
Gut ischemia and hypoxia post severe burn leads to breakdown of intestinal epithelial barrier and enteric bacterial translocation (EBT), resulting in serious complications, such as systemic inflammatory response syndrome, sepsis and multiple organ failure. Cystic fibrosis transmembrane conductance regulator (CFTR)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service