Skip to Content
Merck
All Photos(1)

Key Documents

539279

Sigma-Aldrich

Octenyltrichlorosilane, mixture of isomers

96%

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$533.00

$533.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$533.00

About This Item

Linear Formula:
CH2=CH(CH2)6SiCl3
CAS Number:
Molecular Weight:
245.65
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$533.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

96%

impurities

10-15% internal olefin isomers

refractive index

n20/D 1.458 (lit.)

bp

223-224 °C (lit.)

density

1.07 g/mL at 25 °C (lit.)

SMILES string

Cl[Si](Cl)(Cl)CCCCCCC=C

InChI

1S/C8H15Cl3Si/c1-2-3-4-5-6-7-8-12(9,10)11/h2H,1,3-8H2

InChI key

MFISPHKHJHQREG-UHFFFAOYSA-N

General description

Octenyltrichlorosilane, mixture of isomers (OTTS) is a derivative of trichlorosilane that forms a self-assembled monolayer (SAM) on a variety of substrates. It can be utilized in adhesion promotion and in the modification of surface characteristics.[1]

Application

OTTS forms a SAM on silica coated alumina implants which find potential application in orthopedics as a useful biomaterials.[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

201.0 °F - closed cup

Flash Point(C)

93.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synchrotron radiation stimulated etching of SiO2 thin films with a co contact mask for the area-selective deposition of self-assembled monolayer
Wang C and Urisu T
Japanese Journal of Applied Physics, 42(6S), 4016-4016 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service