Skip to Content
Merck
All Photos(1)

Key Documents

420034

Sigma-Aldrich

Trichloro(phenethyl)silane

95%

Synonym(s):

(2-Phenylethyl)trichlorosilane, Phenethyltrichlorosilane

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$60.80

$60.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$60.80

About This Item

Linear Formula:
C6H5CH2CH2SiCl3
CAS Number:
Molecular Weight:
239.60
Beilstein:
2936059
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$60.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

95%

form

liquid

refractive index

n20/D 1.513 (lit.)

bp

93-96 °C/3 mmHg (lit.)

density

1.234 g/mL at 25 °C (lit.)

SMILES string

Cl[Si](Cl)(Cl)CCc1ccccc1

InChI

1S/C8H9Cl3Si/c9-12(10,11)7-6-8-4-2-1-3-5-8/h1-5H,6-7H2

InChI key

FMYXZXAKZWIOHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trichloro(phenethyl)silane (PTS) is a silane based capping agent that can be used to form a self-assembled monolayer (SAM) on a variety of surfaces. It facilitates the surface treatment and modification for effective detection of surface atoms.[1][2]

Application

A self-assembled monolayer of trichloro(phenethyl)silane on a gate dielectric allows to reduce the transistor channel length.[3]
PTS forms a SAM on a wide range of substrates such as aluminum oxide and silica, which can be further coated on organic field effect transistors (OFETs), thin film transistors and other biosensor based devices for biomedical applications.[4][5][6][7]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Side-chain modulation of dithienofluorene-based copolymers to achieve high field-effect mobilities.
Lee C, et al.
Chemical Science, 8(4), 2942-2951 (2017)
Shifted transfer characteristics of organic thin film and single crystal FETs.
Pernstich KP, et al.
Synthetic Metals, 146(3), 325-328 (2004)
Ultrasensitive silicon nanowire for real-world gas sensing: noninvasive diagnosis of cancer from breath volatolome.
Shehada N, et al.
Nano Letters, 15(2), 1288-1295 (2014)
Organic RFID transponder chip with data rate compatible with electronic product coding.
Organic Electronics, 11(7), 1176-1179 (2010)
Design and realization of a flexible QQVGA AMOLED display with organic TFTs.
Steudel S, et al.
Organic Electronics, 13(9), 1729-1735 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service