Skip to Content
Merck
All Photos(2)

Key Documents

518247

Sigma-Aldrich

Samarium(III) nitrate hexahydrate

99.999% trace metals basis

Synonym(s):

Samarium trinitrate, Trinitric acid samarium(III)·6hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sm(NO3)3 · 6H2O
CAS Number:
Molecular Weight:
444.47
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
crystalline

Assay

99.999% trace metals basis

form

crystalline

reaction suitability

reagent type: catalyst
core: samarium

impurities

≤20 ppm Trace Rare Earth Analysis

SMILES string

[Sm+3].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/3NO3.6H2O.Sm/c3*2-1(3)4;;;;;;;/h;;;6*1H2;/q3*-1;;;;;;;+3

InChI key

HDCOFJGRHQAIPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service