Skip to Content
Merck
All Photos(3)

Key Documents

451134

Sigma-Aldrich

Gadolinium(III) nitrate hexahydrate

99.99% trace metals basis

Synonym(s):

Gadolinium nitrate (Gd(NO)) hexahydrate, Gadolinium trinitrate hexahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$289.00
50 G
$923.00

$289.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$289.00
50 G
$923.00

About This Item

Linear Formula:
Gd(NO3)3 · 6H2O
CAS Number:
Molecular Weight:
451.36
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
solid

$289.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.99% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: gadolinium

impurities

≤150.0 ppm Trace Rare Earth Analysis

mp

91 °C (lit.)

density

2.332 g/mL at 25 °C

SMILES string

[Gd+3].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Gd.3NO3.6H2O/c;3*2-1(3)4;;;;;;/h;;;;6*1H2/q+3;3*-1;;;;;;

InChI key

XWFVFZQEDMDSET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The product was used as a precursor for the preparation of luminescent lanthanide(Ln) nanoparticles for biosensing and imaging.1 It was used to study the pre-concentration and its removal of Gd (III) from laboratory waste aqueous effluent by using a poly hydroxamate (PHA) chelator.[1]

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly-hydroxamic acid (PHA) matrix for gadolinium pre-concentration and removal.
Singha M, et al.
J. Radioanal. Nucl. Chem., 302(2), 961-966 (2014)

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service