Skip to Content
Merck
Samarium(III) nitrate hexahydrate 99.9% trace metals basis

Samarium(III) nitrate hexahydrate

Samarium(III) nitrate hexahydrate 99.9% trace metals basis
Synonym(s):
Samarium trinitrate, Trinitric acid samarium(III)·6hydrate
Linear Formula:
Sm(NO3)3 · 6H2O
CAS Number:
Molecular Weight:
444.47
EC Number:

Products

Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing