Skip to Content
Merck
All Photos(3)

Key Documents

762350

Sigma-Aldrich

Potassium trifluoromethanesulfonimide

≥95%

Synonym(s):

Potassium bis(trifluoromethanesulfonyl)imide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$110.00
5 G
$388.00

$110.00


Estimated to ship onMarch 28, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$110.00
5 G
$388.00

About This Item

Empirical Formula (Hill Notation):
C2F6KNO4S2
CAS Number:
Molecular Weight:
319.24
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
≥95%

$110.00


Estimated to ship onMarch 28, 2025


Request a Bulk Order

Quality Level

Assay

≥95%

form

solid

mp

198-203 °C

SMILES string

[K+].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C2F6NO4S2.K/c3-1(4,5)14(10,11)9-15(12,13)2(6,7)8;/q-1;+1

InChI key

KVFIZLDWRFTUEM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Potassium trifluoromethanesulfonimide is a Lewis acid used to enhance the low-temperature radical polymerization of N, N-dimethylacrylamide to synthesize poly(N,N-dimethylacrylamide).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service