Skip to Content
Merck
All Photos(1)

Documents

51059

Sigma-Aldrich

1-Ethyl-3-methylimidazolium tetrachloroaluminate

≥95%

Synonym(s):

1-Ethyl-3-methyl-1H-imidazolium tetrachloroaluminate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11AlCl4N2
CAS Number:
Molecular Weight:
279.96
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥94.5% (HPLC)
≥95%

refractive index

n20/D 1.484

density

1.304 g/mL at 25 °C

SMILES string

Cl[Al-](Cl)(Cl)Cl.CCn1cc[n+](C)c1

InChI

1S/C6H11N2.Al.4ClH/c1-3-8-5-4-7(2)6-8;;;;;/h4-6H,3H2,1-2H3;;4*1H/q+1;+3;;;;/p-4

InChI key

UYYXEZMYUOVMPT-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

424.4 °F

Flash Point(C)

218 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tingting Wang et al.
Oncology letters, 19(3), 1727-1734 (2020-03-21)
A Disintegrin And Metalloprotease Domain 29 (ADAM29) and Family with sequence similarity 135 member B (FAM135B) genes have been reported to be associated with a carcinogenic risk of esophageal squamous cell carcinoma (ESCC). However, to the best of our knowledge

Articles

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service