Skip to Content
Merck
All Photos(1)

Key Documents

490733

Sigma-Aldrich

Sodium pyruvate-3-13C

99 atom % 13C

Synonym(s):

Pyruvic acid-3-13C sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$898.00
250 MG
$1,200.00
1 G
$1,380.00

$898.00


Estimated to ship onMay 28, 2025


Request more information

Select a Size

Change View
500 MG
$898.00
250 MG
$1,200.00
1 G
$1,380.00

About This Item

Linear Formula:
13CH3COCO2Na
CAS Number:
Molecular Weight:
111.04
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

$898.00


Estimated to ship onMay 28, 2025


Request more information

isotopic purity

99 atom % 13C

Quality Level

Assay

99% (CP)

form

solid

technique(s)

bio NMR: suitable
mass spectrometry (MS): suitable

mp

>300 °C (lit.)

mass shift

M+1

storage temp.

2-8°C

SMILES string

[Na+].[13CH3]C(=O)C([O-])=O

InChI

1S/C3H4O3.Na/c1-2(4)3(5)6;/h1H3,(H,5,6);/q;+1/p-1/i1+1;

InChI key

DAEPDZWVDSPTHF-YTBWXGASSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium pyruvate-3-13C is an isotopic analogue of sodium pyruvate.

Application

Sodium pyruvate-3-13C is used to study the brain metabolism of exogenous pyruvate.It is also used to investigate the pyruvate metabolism in living cancer cells.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susana Villa Gonzalez et al.
Journal of neurochemistry, 95(1), 284-293 (2005-09-27)
Pyruvate given in large doses may be neuroprotective in stroke, but it is not known to what degree the brain metabolizes pyruvate. Intravenous injection of [3-13C]pyruvate led to dose-dependent labelling of cerebral metabolites so that at 5 min after injection
Chendong Yang et al.
The Journal of biological chemistry, 289(9), 6212-6224 (2014-01-15)
Metabolic reprogramming facilitates cancer cell growth, so quantitative metabolic flux measurements could produce useful biomarkers. However, current methods to analyze flux in vivo provide either a steady-state overview of relative activities (infusion of (13)C and analysis of extracted metabolites) or

Articles

Isotopic Labeling for NMR Spectroscopy of Biological Solids;

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service