Skip to Content
Merck
All Photos(3)

Key Documents

746258

Sigma-Aldrich

L-Lactic acid-13C3

greener alternative

≥99 atom % 13C, ≥98% (CP), ≥98% (Chiral Purity, HPLC)

Synonym(s):

13C Labeled Lactic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$967.00
1 G
$11,670.00

$967.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
$967.00
1 G
$11,670.00

About This Item

Empirical Formula (Hill Notation):
13C3H6O3
CAS Number:
Molecular Weight:
93.06
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

$967.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

≥99 atom % 13C

Quality Level

Assay

≥98% (CP)
≥98% (Chiral Purity, HPLC)

form

solid

greener alternative product score

old score: 45
new score: 14
Find out more about DOZN™ Scoring

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Design for Energy Efficiency
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

bio NMR: suitable
mass spectrometry (MS): suitable

greener alternative category

mass shift

M+3

shipped in

dry ice

storage temp.

−20°C

SMILES string

[13CH3][13C@H](O)[13C](O)=O

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)/t2-/m0/s1/i1+1,2+1,3+1

InChI key

JVTAAEKCZFNVCJ-GCCOVPGMSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses”, “Design for Energy Efficiency” and “Inherently Safer Chemistry for Accident Prevention”. Click here to view its DOZN scorecard.

Application

L-(+)-Lactic Acid-13C3, a stable isotope compound, is used in the investigation of human lactate metabolism.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R A Neese et al.
Biomedical mass spectrometry, 10(8), 458-462 (1983-08-01)
A stable isotope tracer method has been developed for studying lactate metabolism in humans. The method uses lactic acid triple labeled with 13C as the tracer. The stable isotope is infused to attain a level of approximately 1.5% of that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service