Skip to Content
Merck
All Photos(1)

Documents

704334

Sigma-Aldrich

2-Ketoglutaric acid-1-13C

99 atom % 13C, 95% (CP)

Synonym(s):

α-Ketoglutaric acid-1-13C, 2-Ketopentanedioic acid-1-13C, 2-Oxoglutaric acid-1-13C, 2-Oxopentanedioic acid-1-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCOCH2CH2CO13CO2H
CAS Number:
Molecular Weight:
147.09
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

95% (CP)

form

solid

technique(s)

bio NMR: suitable

mp

114-117 °C

mass shift

M+1

storage temp.

2-8°C

SMILES string

OC(=O)CCC(=O)[13C](O)=O

InChI

1S/C5H6O5/c6-3(5(9)10)1-2-4(7)8/h1-2H2,(H,7,8)(H,9,10)/i5+1

InChI key

KPGXRSRHYNQIFN-HOSYLAQJSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service