Skip to Content
Merck
All Photos(1)

Key Documents

490431

Sigma-Aldrich

Oleic acid-13C18

≥99 atom % 13C, ≥99% (CP)

Synonym(s):

13C Labeled oleic acid, Elainic acid-13C18, Oleic acid-ul-13C

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
Inquire
1 G
Inquire

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
Inquire
1 G
Inquire

About This Item

Linear Formula:
13CH3(13CH2)713CH=13CH(13CH2)713CO2H
CAS Number:
Molecular Weight:
300.33
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.12

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

grade

analytical standard

isotopic purity

≥99 atom % 13C

Assay

≥99% (CP)

form

liquid

refractive index

n20/D 1.4595 (lit.)

bp

192-195 °C/1.2 mmHg (lit.)

mp

13.4 °C (lit.)

density

0.947 g/mL at 25 °C

format

neat

mass shift

M+18

storage temp.

−20°C

SMILES string

[H]\[13C]([13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH3])=[13C](/[H])[13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13C](O)=O

InChI

1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H,19,20)/b10-9-/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1,13+1,14+1,15+1,16+1,17+1,18+1

InChI key

ZQPPMHVWECSIRJ-IGBBIXMTSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Oleic acid-13C18 is a 13C labeled fatty acid. The disposition of 13C18 -oleic acid following in vivo intravenous and oral administration has been determined by liquid chromatography-mass spectrometry.[1]

Application

Oleic acid-13C18 was used to prepare stable isotope-labeled fatty acid (FA) mixtures.[2] It was also used for the isotopic labeling of triglycerides (TG) in HepG2 hepatoma cells.[3]

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Quality

This product may be used as an analytical standard.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiqi Zhang et al.
Journal of animal science and biotechnology, 5(1), 29-29 (2014-07-02)
Fatty acid (FA) composition is the most important parameter affecting the flavor and nutritional value of the meat. The final and the only committed step in the biosynthesis of triglycerides is catalyzed by diacylglycerol acyltransferase 2 (DGAT2). The role of
Jenson Qi et al.
Journal of lipid research, 53(6), 1106-1116 (2012-04-12)
Diacylglycerol acyltransferase (DGAT) catalyzes the final step in triglyceride (TG) synthesis. There are two isoforms, DGAT1 and DGAT2, with distinct protein sequences and potentially different physiological functions. To date, the ability to determine clear functional differences between DGAT1 and DGAT2
David G McLaren et al.
Analytical chemistry, 85(13), 6287-6294 (2013-05-15)
We have previously reported on a liquid chromatography-mass spectrometry method to determine the disposition of [(13)C18]-oleic acid following intravenous and oral administration in vivo. This approach has enabled us to study a variety of aspects of lipid metabolism including a
Soudabeh Imanikia et al.
Cell reports, 28(3), 581-589 (2019-07-18)
The endoplasmic reticulum unfolded protein response (UPRER) is a cellular stress response that maintains homeostasis within the secretory pathway, regulates glucose and lipid metabolism, and influences longevity. To ask whether this role in lifespan determination depends upon metabolic intermediaries, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service