Skip to Content
Merck
All Photos(1)

Key Documents

299162

Sigma-Aldrich

Stearic acid-1-13C

99 atom % 13C

Synonym(s):

1-Heptadecanecarboxylic acid-1-13C, 13C Labeled stearic acid, Cetylacetic acid-1-13C, Octadecanoic acid-1-13C, Stearophanic acid-1-13C

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$832.00

$832.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$832.00

About This Item

Linear Formula:
CH3(CH2)1613CO2H
CAS Number:
Molecular Weight:
285.47
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

$832.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C

Quality Level

Assay

99% (CP)

form

solid

bp

361 °C (lit.)

mp

68-70 °C (lit.)

mass shift

M+1

SMILES string

CCCCCCCCCCCCCCCCC[13C](O)=O

InChI

1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)/i18+1

InChI key

QIQXTHQIDYTFRH-CPZJZEHKSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Zou et al.
Clinica chimica acta; international journal of clinical chemistry, 483, 164-169 (2018-05-02)
No index for non-invasive diagnosis of subclinical pelvic inflammatory disease (PID) is available at this time. Here we carried out a plasma metabolomic study to search for potential biomarkers to facilitate its non-invasive diagnosis. The metabolites in plasma were detected
Mike Lange et al.
Cell reports. Medicine, 2(10), 100407-100407 (2021-11-11)
Obesity, characterized by expansion and metabolic dysregulation of white adipose tissue (WAT), has reached pandemic proportions and acts as a primer for a wide range of metabolic disorders. Remodeling of WAT lipidome in obesity and associated comorbidities can explain disease

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service