489166
Lauric-2,2-d2 acid
98 atom % D
Synonym(s):
Dodecanoic acid-2,2-d2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
isotopic purity
98 atom % D
Quality Level
form
solid
bp
225 °C/100 mmHg (lit.)
mp
44-46 °C (lit.)
mass shift
M+2
SMILES string
[2H]C([2H])(CCCCCCCCCC)C(O)=O
InChI
1S/C12H24O2/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2-11H2,1H3,(H,13,14)/i11D2
InChI key
POULHZVOKOAJMA-ZWGOZCLVSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
>235.4 °F
Flash Point(C)
> 113.00 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service