Skip to Content
Merck
All Photos(1)

Documents

490938

Sigma-Aldrich

Sodium trifluoroacetate-1-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF313CO2Na
CAS Number:
Molecular Weight:
137.00
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Quality Level

form

solid

mp

207 °C (dec.) (lit.)

mass shift

M+1

SMILES string

[Na+].[O-][13C](=O)C(F)(F)F

InChI

1S/C2HF3O2.Na/c3-2(4,5)1(6)7;/h(H,6,7);/q;+1/p-1/i1+1;

InChI key

UYCAUPASBSROMS-YTBWXGASSA-M

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service