Skip to Content
Merck
All Photos(1)

Key Documents

464503

Sigma-Aldrich

2-Decyl-1-tetradecanol

97%

Synonym(s):

2-Decyl-1-tetradecanol, 2-Decylmyristyl alcohol, 2-Decyltetradecanol, 2-Decyltetradecyl alcohol, 2-Dodecyl-1-dodecanol, Decyltetradecanol, Decyltetradecyl alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$149.00

$149.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$149.00

About This Item

Linear Formula:
CH3(CH2)11CH[(CH2)9CH3]CH2OH
CAS Number:
Molecular Weight:
354.65
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$149.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.457 (lit.)

bp

271-275 °C/33 mmHg (lit.)

mp

17-20 °C (lit.)

density

0.842 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCCCCCCCCCCCC(CO)CCCCCCCCCC

InChI

1S/C24H50O/c1-3-5-7-9-11-13-14-16-18-20-22-24(23-25)21-19-17-15-12-10-8-6-4-2/h24-25H,3-23H2,1-2H3

InChI key

CAYHVMBQBLYQMT-UHFFFAOYSA-N

General description

2-Decyl-1-tetradecanol participates in the preparation of monosaccharide glycolipid.[1]

Application

2-Decyl-1-tetradecanol may be used in the preparation of 2-decyltetradecyl (4-nitrophenyl)[11-([(2,5-dioxotetrahydro-1H-1-pyrrolyl)oxy]carbonyloxy)undecyl]phosphonate.[2]

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

410.0 °F - open cup

Flash Point(C)

210 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H J Deussen et al.
Bioorganic & medicinal chemistry letters, 10(17), 2027-2031 (2000-09-15)
The design, synthesis, and inhibition properties of two new triglyceride analogue biotinylated suicide inhibitors (2) and (3) for directed molecular evolution of lipolytic enzymes by phage-display is described.
Specific surface modification of the acetylene-linked glycolipid vesicle by click chemistry.
Ito H, et al.
Chemical Communications (Cambridge, England), 48(45), 5650-5652 (2012)
Andrea Garcia-Ortiz et al.
ChemSusChem, 13(4), 707-714 (2020-01-09)
Biomass-derived surfactants with very good surface tension and critical micellar concentration properties were obtained by conversion of methyl levulinate into methyl 4-alkoxypentanoates through reductive etherification with aliphatic alcohols. Among different bifunctional acid/metal catalysts best results were obtained with Pd on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service