Skip to Content
Merck
All Photos(3)

Key Documents

249416

Sigma-Aldrich

2-Ethylhexyl bromide

contains 1% potassium carbonate as stabilizer, 95%

Synonym(s):

1-Bromo-2-ethylhexane

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$57.60
500 G
$184.00

$57.60


Estimated to ship onMarch 23, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$57.60
500 G
$184.00

About This Item

Linear Formula:
CH3(CH2)3CH(C2H5)CH2Br
CAS Number:
Molecular Weight:
193.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$57.60


Estimated to ship onMarch 23, 2025


Request a Bulk Order

Quality Level

Assay

95%

form

liquid

contains

1% potassium carbonate as stabilizer

refractive index

n20/D 1.4538 (lit.)

bp

75-77 °C/16 mmHg (lit.)

density

1.086 g/mL at 25 °C (lit.)

functional group

alkyl halide
bromo

storage temp.

2-8°C

SMILES string

CCCCC(CC)CBr

InChI

1S/C8H17Br/c1-3-5-6-8(4-2)7-9/h8H,3-7H2,1-2H3

InChI key

NZWIYPLSXWYKLH-UHFFFAOYSA-N

Application

2-Ethylhexyl bromide has been used in the synthesis of:
  • methoxy-4-(2-ethylhexyloxy)benzene[1]
  • photovoltaic conjugated polymer, having benzo[1,2-b:4,5-b′]difuran building block[2]
  • 1-bromo-4-(2′-ethylhexyloxy)benzene3

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lijun Huo et al.
Chemical communications (Cambridge, England), 48(27), 3318-3320 (2012-03-01)
A new building block of benzo[1,2-b:4,5-b']difuran (BDF) was firstly designed and synthesized. The newly designed unit was applied for constructing a new photovoltaic low band gap polymer, PBDFDTBT, which exhibited promising power conversion efficiency of 5.0%.
MEH-PPV: improved synthetic procedure and molecular weight control.
Neef CJ and Ferraris JP.
Macromolecules, 33(7), 2311-2314 (2000)
Siva Doddi et al.
Journal of fluorescence, 26(6), 1939-1949 (2016-11-01)
We report the synthesis and characterization of a new fluorescent dyad SP-DPP-SP(9) via efficient palladium-catalyzed Sonogashira coupling of prop-2-yn-1-yl 3-(3',3'dimethyl-6-nitrospiro[chromene-2,2'-indolin]-1'-yl)propanoatespiropyran, SP(8), a well known photochromic accepter, with 3,6-bis(5-bromothiophen-2-yl)-2,5-bis((R)-2-ethylhexyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione, DPP(4), a highly fluorescent donor. Under visible light exposure the SP unit
Efficient 9-alkylphenyl-9-pyrenylfluorene substituted pyrene derivatives with improved hole injection for blue light-emitting diodes.
Tang C, et al.
Journal of Materials Chemistry, 16(41), 4074-4080 (2006)
Marcelo D T Torres et al.
ACS nano, 15(1), 966-978 (2021-01-14)
Antibiotic-resistant bacteria infect close to 3 million people, and kill 35,000, each year in the United States. Ionic liquid (IL)-based antimicrobial agents have the potential to diversify our ever-diminishing antibiotic arsenal. Here, we describe an IL with potent submicromolar antimicrobial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service