Skip to Content
Merck
All Photos(1)

Key Documents

364622

Sigma-Aldrich

Phenanthrene-d10

98 atom % D, 98% (CP)

Synonym(s):

Perdeuterated phenanthrene

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$343.00

$343.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
$343.00

About This Item

Empirical Formula (Hill Notation):
C14D10
CAS Number:
Molecular Weight:
188.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.12

$343.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

Assay

98% (CP)

form

solid

bp

340 °C (lit.)

mp

98-100 °C (lit.)

application(s)

electronics

mass shift

M+10

storage temp.

room temp

SMILES string

[2H]c1c([2H])c([2H])c2c(c1[2H])c([2H])c([2H])c3c([2H])c([2H])c([2H])c([2H])c23

InChI

1S/C14H10/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)13/h1-10H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D

InChI key

YNPNZTXNASCQKK-LHNTUAQVSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

May be used as an analytical standard

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ayah Alassali et al.
Polymers, 12(6) (2020-06-19)
Due to the lack of advanced methods to clean plastic waste from organic contaminants, this study aimed at evaluating supercritical extraction as a decontamination method. Oil-adhesive high-density polyethylene (HD-PE) oil containers were subjected to supercritical extraction using supercritical carbon dioxide.
Ge Li et al.
Food chemistry, 209, 123-130 (2016-05-14)
This study investigated the effects of frying conditions on the change, migration and correlation of polycyclic aromatic hydrocarbons (PAHs) and oxygenated PAHs (oxy-PAHs) in youtiao, a typical Chinese fried bread. PAH migration was evaluated using an isotope tracer method. High
Célia A Alves et al.
Environmental science and pollution research international, 22(15), 11526-11542 (2015-04-02)
The main purpose of this work was to evaluate the chemical composition of particulate matter (PM) emitted by eight different light-duty vehicles. Exhaust samples from petrol and diesel cars (Euro 3 to Euro 5) were collected in a chassis dynamometer
Dennis S Mkhize et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 79(5), 808-819 (2019-04-27)
A passive sampling device, based on molecularly imprinted membranes (MIM), was fabricated and optimised for sampling polychlorinated biphenyls (PCBs) in aquatic ecosystems. The newly-developed passive sampler was subjected to in-situ calibration studies to determine PCB sampling rates under various conditions
Wan-Ting Chang et al.
Marine pollution bulletin, 97(1-2), 217-223 (2015-06-16)
This work investigates the dependence of the sampling rate (Rs) of semi-permeable membrane devices (SPMDs) on flow rate and temperature. The in situ Rs values were obtained using performance reference compounds (PRCs) with weighted polynomial regression and used to estimate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service