Skip to Content
Merck
All Photos(1)

Key Documents

449059

Sigma-Aldrich

4-(Octyloxy)benzonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7OC6H4CN
CAS Number:
Molecular Weight:
231.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.513 (lit.)

bp

171-173 °C/2 mmHg (lit.)

density

0.967 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCOc1ccc(cc1)C#N

InChI

1S/C15H21NO/c1-2-3-4-5-6-7-12-17-15-10-8-14(13-16)9-11-15/h8-11H,2-7,12H2,1H3

InChI key

GFNSBTARZPEIPN-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Georg M Fischer et al.
    Chemistry (Weinheim an der Bergstrasse, Germany), 15(19), 4857-4864 (2009-03-20)
    Pyrrolopyrrole cyanine (PPCy) dyes are presented as a novel class of near-infrared (NIR) chromophores, which are synthesized in a condensation reaction of diketopyrrolopyrrole with heteroarylacetonitrile compounds. Their optical properties are marked by strong and narrow-band NIR absorptions. Complexation products with
    Synthesis of liquid crystals materials derived from oxadiazole, isoxazole and tetrazole heterocycles.
    dos Santos DR, et al.
    ARKIVOC (Gainesville, FL, United States), 17, 157-166 (2008)

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service