Skip to Content
Merck
All Photos(1)

Key Documents

SML2530

Sigma-Aldrich

CAY10526

≥98% (HPLC)

Synonym(s):

4-(Benzo[b]thiophen-2-yl)-3-bromo-5-hydroxyfuran-2(5H)-one, 4-Benzo[b]thien-2-yl-3-bromo-5-hydroxy-2(5H)-furanone, 4-Benzo[b]thiophen-2-yl-3-bromo-5-hydroxy-5H-furan-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$237.00
25 MG
$750.00

$237.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$237.00
25 MG
$750.00

About This Item

Empirical Formula (Hill Notation):
C12H7BrO3S
CAS Number:
Molecular Weight:
311.15
UNSPSC Code:
51111800
NACRES:
NA.77

$237.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1OC(C(C2=CC3=C(S2)C=CC=C3)=C1Br)O

InChI

1S/C12H7BrO3S/c13-10-9(11(14)16-12(10)15)8-5-6-3-1-2-4-7(6)17-8/h1-5,11,14H

InChI key

ICCDILPNCJIDAW-UHFFFAOYSA-N

Biochem/physiol Actions

CAY10526 is a selective inhibitor of microsomal prostaglandin E2 synthase-1 (mPGES-1) expression. CAY10526 potently inhibits PGE2 production in lipopolysaccharide-stimulated RAW 264.7 cells. CAY10526 significantly suppressed tumor growth and increased apoptosis in melanoma xenografts.
selective inhibitor of microsomal prostaglandin E2 synthase-1 (mPGES-1) expression that potently inhibits PGE2 production

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Darren J Perkins et al.
Nature immunology, 19(12), 1309-1318 (2018-11-07)
The unique cell biology of Toll-like receptor 4 (TLR4) allows it to initiate two signal-transduction cascades: a signal dependent on the adaptors TIRAP (Mal) and MyD88 that begins at the cell surface and regulates proinflammatory cytokines, and a signal dependent
Sun-Hee Kim et al.
Pigment cell & melanoma research, 29(3), 297-308 (2016-01-24)
COX-2 and its product PGE2 enhance carcinogenesis and tumor progression, which has been previously reported in melanoma. As most COX inhibitors cause much toxicity, the downstream microsomal PGE2 synthase-1 (mPGES1) is a consideration for targeting. Human melanoma TMAs were employed
Maria D Guerrero et al.
Journal of medicinal chemistry, 50(9), 2176-2184 (2007-04-05)
As a part of our drug discovery effort, recently we clarified the molecular basis of phospholipase A2 (PLA2) inactivation by petrosaspongiolide M (PM), an interesting metabolite belonging to a marine sesterterpene family, containing in its structural architecture a gamma-hydroxybutenolide moiety

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service