Skip to Content
Merck
All Photos(1)

Key Documents

436291

Sigma-Aldrich

Octylmagnesium bromide solution

2.0 M in diethyl ether

Synonym(s):

1-Octylmagnesium bromide, n-Octylmagnesium bromide, Bromooctylmagnesium

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$127.00
800 ML
$512.00

$127.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$127.00
800 ML
$512.00

About This Item

Linear Formula:
CH3(CH2)7MgBr
CAS Number:
Molecular Weight:
217.43
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$127.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

reaction suitability

reaction type: Grignard Reaction

concentration

2.0 M in diethyl ether

bp

34.6 °C

density

0.961 g/mL at 25 °C

SMILES string

CCCCCCCC[Mg]Br

InChI

1S/C8H17.BrH.Mg/c1-3-5-7-8-6-4-2;;/h1,3-8H2,2H3;1H;/q;;+1/p-1

InChI key

ZDUZWGUHIRQPGP-UHFFFAOYSA-M

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-31.0 °F - closed cup

Flash Point(C)

-35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Verónica Fernández-Stefanuto et al.
Frontiers in chemistry, 8, 208-208 (2020-04-09)
Surfactants have a great number of applications. Among these chemicals, petroleum sulfonates have been widely used due to their effectiveness in reducing interfacial tension. This is the case of sodium octylnaphthalene sulfonate which is a solid with a very low

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service