Skip to Content
Merck
All Photos(1)

Key Documents

637963

Sigma-Aldrich

3,7-Dimethyloctylmagnesium bromide solution

1.0 M in diethyl ether

Synonym(s):

Bromo(3,7-dimethyloctyl)magnesium

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$647.00

$647.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$647.00

About This Item

Empirical Formula (Hill Notation):
C10H21BrMg
CAS Number:
Molecular Weight:
245.48
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$647.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in diethyl ether

density

0.856 g/mL at 25 °C

SMILES string

CC(C)CCCC(C)CC[Mg]Br

InChI

1S/C10H21.BrH.Mg/c1-5-10(4)8-6-7-9(2)3;;/h9-10H,1,5-8H2,2-4H3;1H;/q;;+1/p-1

InChI key

CZIGPVDHJSGGIU-UHFFFAOYSA-M

Application


  • Evaluating how cationic lipid affects mRNA-LNP physical properties and biodistribution: This study describes the synthesis process where 3,7-Dimethyloctylmagnesium bromide was used as a reagent in the preparation of a lipid component for lipid nanoparticles (LNPs), which are crucial in mRNA delivery systems (Gueguen et al., 2024).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-13.0 °F - closed cup

Flash Point(C)

-25 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service