Skip to Content
Merck
All Photos(2)

Key Documents

429457

Sigma-Aldrich

Lithium chloride

greener alternative

AnhydroBeads, −10 mesh, 99.998% trace metals basis

Synonym(s):

Lithium monochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiCl
CAS Number:
Molecular Weight:
42.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for synthesis
synthesis grade
form:
beads
solubility:
H2O: soluble
Pricing and availability is not currently available.

grade

for synthesis
synthesis grade

product line

AnhydroBeads

Assay

99.998% trace metals basis

form

beads

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤25.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

605 °C (lit.)

solubility

H2O: soluble

greener alternative category

SMILES string

[Li+].[Cl-]

InChI

1S/ClH.Li/h1H;/q;+1/p-1

InChI key

KWGKDLIKAYFUFQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Lithium chloride, an anhydrous lithiumsalt, is highly soluble in polar solvents like water and exhibits hygroscopicproperties. LiCl can be used as an electrolyte additive in lithium-ion batterytechnology to improve the performance and stability of lithium-ion batteries. Itis also used in several applications in the field of materials scienceincluding, the production of lithium metal, desiccant, solid-state electrolytes, gassensors, catalysis, and organic synthesis.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Lithium chloride can be used as:
  • A molten salt electrolyte in Li-based liquid metal batteries. LiCl facilitates the transport of lithium ions between the electrodes during the battery′s charge and discharge cycles. LiCl, along with KCl, contributes to the ionic conductivity and stability of the molten salt electrolyte.
  • A precursor material for the development of fluorine-substituted lithium chloride solid electrolytes for high-voltage solid-state lithium-ion batteries.
  • A salt additive in the formulation of a gel electrolyte for quasi-solid dye-sensitized solar cells. LiCl enhances the ionic conductivity of the gel electrolyte, facilitating the movement of charges within the solar cell
  • An additive in the development of a composite electron transport layer for N-I-P type monolithic perovskite/silicon tandem solar cells. It is added to the tin oxide (SnO2) precursor solution to passivate defects within the SnO2 layer and enhance the electron injection driving force at the interface between the electron transfer layer (ETL) and perovskite.
  • Molten salts reactors as an coolant

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoqi Sun et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 3(8), 1600044-1600044 (2016-11-08)
The major advantage of Mg batteries relies on their promise of employing an Mg metal negative electrode, which offers much higher energy density compared to graphitic carbon. However, the strong coulombic interaction of Mg
Tang, J. et al.
Tetrahedron, 55, 1893-1893 (1999)
Alessandro Di Maio et al.
EvoDevo, 6, 17-17 (2015-07-15)
Wnt signaling is one of the earliest and most highly conserved regulatory pathways for the establishment of the body axes during regeneration and early development. In regeneration, body axes determination occurs independently of tissue rearrangement and early developmental cues. Modulation
Aitor González et al.
PloS one, 8(1), e53323-e53323 (2013-01-18)
The mouse segmentation is established from somites, which are iteratively induced every two hours from the presomitic mesoderm (PSM) by a system known as the segmentation clock. A crucial component of the segmentation clock is the gene Hes7, which is
Karla M Acevedo et al.
The Journal of biological chemistry, 289(16), 11007-11019 (2014-03-13)
Amyloid precursor protein (APP) undergoes post-translational modification, including O- and N-glycosylation, ubiquitination, and phosphorylation as it traffics through the secretory pathway. We have previously reported that copper promotes a change in the cellular localization of APP. We now report that

Articles

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service