Skip to Content
Merck
All Photos(3)

Key Documents

357405

Sigma-Aldrich

Zirconium(IV) chloride

≥99.9% trace metals basis

Synonym(s):

Tetrachlorozirconium, Zirconium tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZrCl4
CAS Number:
Molecular Weight:
233.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.9% trace metals basis
form:
powder

vapor pressure

1 mmHg ( 190 °C)

Quality Level

Assay

≥99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: zirconium

impurities

≤1000.0 ppm Trace Metal Analysis

transition temp

sublimation point 331 °C

density

2.8 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

Cl[Zr](Cl)(Cl)Cl

InChI

1S/4ClH.Zr/h4*1H;/q;;;;+4/p-4

InChI key

DUNKXUFBGCUVQW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Zirconium (IV) chloride acts as an efficient Lewis acid catalyst for various reactions; such as:
  • electrophilic addition reaction of indole with aldehydes/ketones to form bis(indolyl)methanes. electrophilic amination of activated arenes,
  • transthioacetylization of acetals,
  • deoxygenation of heterocyclic-N-oxides,
  • reduction of nitro compounds,
  • conversion of carbonyl compounds to 1,3-oxathiolanes,
  • Biginelli reaction for the synthesis of 3,4-dihydropyrimidin-2(1H)-ones,
  • direct amide coupling of non activated carboxylic acids and amines,
  • synthesis of quinoxalines and pyrido[2,3-b]pyrazines,
  • deprotection of t-butyldimethylsilyl (TBDMS) ethers.
Activates pyrrolidines for improved conversion, via a modified Bouveault reaction, to the corresponding α,α-dimethylamines.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M
Concise Encyclopedia Chemistry null
Zirconium(IV) Chloride - Catalysed Reaction of Indoles: An Expeditious Synthesis of Bis(indolyl)methanes
Nagawade RR, et al.
Bull. Korean Chem. Soc., 26(12) (2005)
Aldrichimica Acta, 18, 31-31 (1985)
Zirconium(IV) chloride as versatile catalyst for the expeditious synthesis of quinoxalines and pyrido[2,3-b]pyrazines under ambient conditions
Aghapoor K, et al.
Transition Metal Chemistry, 35(1), 49-53 null
A facile zirconium(IV) chloride catalysed selective deprotection of t-butyldimethylsilyl (TBDMS) ethers
Sharma GVM, et al.
Tetrahedron Letters, 44, 4689-4691 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service