Skip to Content
Merck
All Photos(1)

Key Documents

1.05679

Supelco

Lithium chloride

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Lithium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
ClLi
CAS Number:
Molecular Weight:
42.39
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-212-3
NACRES:
NA.21
grade:
ACS reagent
form:
solid
solubility:
832 g/L

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

vapor pressure

1.33 hPa ( 547 °C)

product line

EMSURE®

Assay

≥99% (LiCl basis, silver nitrate titration)

form

solid

potency

526 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

impurities

≤0.008 meq/g Total of alkali
≤0.01% Insoluble matter

loss

≤1% loss on drying, 130 °C

pH

6 (20 °C, 50 g/L in H2O)

bp

1360 °C/1013 hPa

mp

614 °C

solubility

832 g/L

density

2.07 g/cm3 at 20 °C

bulk density

530 kg/m3

anion traces

nitrate (NO3-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Ba: ≤0.002%
Ca: ≤0.005%
Fe: ≤0.0005%
K: ≤0.01%
Mg: ≤0.005%
Na: ≤0.02%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[Li+].[Cl-]

InChI

1S/ClH.Li/h1H;/q;+1/p-1

InChI key

KWGKDLIKAYFUFQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Stereocontrolled synthesis of syn-beta-Hydroxy-alpha-amino acids by direct aldolization of pseudoephenamine glycinamide.: Lithium chloride is employed in the stereocontrolled synthesis of syn-beta-Hydroxy-alpha-amino acids, highlighting its role in advanced synthetic chemistry techniques. This research provides insights into chemical synthesis and the development of complex organic compounds, relevant for chemists focusing on synthetic methodologies (Seiple et al., 2014).

Analysis Note

Assay (argentometric, LiCl): ≥ 99 %
Insoluble matter: ≤ 0.01 %
Nitrate (NO₃): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.0005 %
Ba (Barium): ≤ 0.002 %
Ca (Calcium): ≤ 0.005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.01 %
Mg (Magnesium): ≤ 0.005 %
Na (Sodium): ≤ 0.02 %
Total of alkali: ≤ 0.008 meq/g
Loss on drying (130 °C): ≤ 1.0 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service