Skip to Content
Merck
All Photos(1)

Documents

420476

Sigma-Aldrich

2,2,2-Trifluoro-2′,4′,6′-trimethylacetophenone

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2COCF3
CAS Number:
Molecular Weight:
216.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97%

form

liquid

refractive index

n20/D 1.456 (lit.)

bp

70-80 °C/20 mmHg (lit.)

density

1.136 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(C)c(c(C)c1)C(=O)C(F)(F)F

InChI

1S/C11H11F3O/c1-6-4-7(2)9(8(3)5-6)10(15)11(12,13)14/h4-5H,1-3H3

InChI key

VINRTVDNUHIWCB-UHFFFAOYSA-N

Application

2,2,2-Trifluoro-2′,4′,6′-trimethylacetophenone may be used for organic synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service