Skip to Content
Merck
All Photos(1)

Key Documents

72248

Sigma-Aldrich

Nickel(II) carbonate basic hydrate

technical, ~46% Ni basis (KT)

Synonym(s):

Basic nickel carbonate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$86.30
1 KG
$167.00

$86.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$86.30
1 KG
$167.00

About This Item

Linear Formula:
~NiCO3 · 2Ni(OH)2 · 4H2O
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$86.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

grade

technical

form

powder

reaction suitability

core: nickel
reagent type: catalyst

concentration

~46% Ni (KT)

impurities

~5% sodium sulfate

SMILES string

[Ni++].O[Ni]O.O[Ni]O.[O-]C([O-])=O

InChI

1S/CH2O3.3Ni.4H2O/c2-1(3)4;;;;;;;/h(H2,2,3,4);;;;4*1H2/q;3*+2;;;;/p-6

InChI key

ATYNVYNPWGEMMI-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service