Skip to Content
Merck
All Photos(2)

Documents

371254

Sigma-Aldrich

Iron(III) oxide

hydrated, catalyst grade, 30-50 mesh

Synonym(s):

Ferric hydroxide oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe(OH)O
CAS Number:
Molecular Weight:
88.85
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

grade

catalyst grade

form

powder

particle size

30-50 mesh

application(s)

battery manufacturing

SMILES string

O[Fe]=O

InChI

1S/Fe.H2O.O/h;1H2;/q+1;;/p-1

InChI key

AEIXRCIKZIZYPM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keunhong Jeong et al.
Magnetic resonance in chemistry : MRC, 56(11), 1089-1093 (2018-06-02)
Parahydrogen is a potentially significant source of hyperpolarization. However, a heat exchanger at an ultralow temperature, which is normally sustained wastefully using liquid nitrogen, is essential for the generation of hyperpolarized parahydrogen. In order to cut down on the use
P A Adhikari et al.
Journal of animal science, 93(1), 209-216 (2015-01-09)
The aim was to determine the true total tract digestibility (TTTD) and standardized total tract digestibility (STTD) of P in canola meals from Brassica napus black (BNB) and Brassica juncea yellow (BJY) fed to growing pigs. Fifty-four barrows with an
Mina Ibrahim Tadros et al.
International journal of pharmaceutics, 472(1-2), 27-39 (2014-06-15)
The current work aimed to develop novel composite sponges of chitosan (CH)-chondroitin sulfate (CS) as a low-density gastroretentive delivery system for lornoxicam (LOR). This triple anti-inflammatory therapy-loaded matrices are expected to expand and float upon contact with gastric fluids for
Jang Han Lee et al.
Environmental toxicology and chemistry, 33(12), 2759-2766 (2014-09-02)
Variable sizes of nanoparticles, ranging from nano to micro scale, are of toxicological interest. In the present study, the authors hypothesized that, in addition to the size, the shape of iron oxide (Fe2O3) nanoparticles is a major factor that contributes

Articles

Magnetic materials find diverse applications from data storage to renewable energy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service