Skip to Content
Merck
All Photos(2)

Key Documents

419869

Sigma-Aldrich

Ethynyl p-tolyl sulfone

98%

Synonym(s):

p-Toluenesulfonylacetylene, Tosylacetylene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$142.00

$142.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$142.00

About This Item

Linear Formula:
CH3C6H4SO2C≡CH
CAS Number:
Molecular Weight:
180.22
Beilstein:
2556169
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$142.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Assay

98%

mp

73-74 °C (lit.)

solubility

organic solvents: soluble(lit.)

functional group

sulfone

SMILES string

Cc1ccc(cc1)S(=O)(=O)C#C

InChI

1S/C9H8O2S/c1-3-12(10,11)9-6-4-8(2)5-7-9/h1,4-7H,2H3

InChI key

FTHKWIMQNXVEHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethynyl p-tolyl sulfone (Tosylacetylene) may be used in the synthesis of 2-(4-methylphenylsulfonyl)ethenyl (tosvinyl, Tsv) protected derivatives.[1] It may be used as starting reagent for the synthesis of optically active indan-2-ols.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena Petit et al.
The Journal of organic chemistry, 79(18), 8826-8834 (2014-08-28)
The use of the 2-(4-methylphenylsulfonyl)ethenyl (tosvinyl, Tsv) group for the protection of the NH group of a series of imides, azinones (including AZT), inosines, and cyclic sulfonamides has been examined. The Tsv-protected derivatives are obtained in excellent yields by conjugate
Takeshi Hanazawa et al.
The Journal of organic chemistry, 68(12), 4980-4983 (2003-06-07)
An efficient and practical synthesis of optically active indan-2-ols 1 has been developed starting from readily accessible optically active 4-siloxy-1,6-alkadiynes 2 and ethynyl p-tolyl sulfone, where the metalative Reppe reaction mediated by an economical divalent titanium reagent, Ti(O-i-Pr)(4)/2 i-PrMgCl, is
A Simplified Method for the Preparation of Ethynyl P-Tolyl Sulfone and Ethynyl Phenyl Sulfone.
Chen Z and Trudell ML.
Synthetic Communications, 24(21), 3149-3155 (1994)
Hao Chen et al.
Bioorganic & medicinal chemistry letters, 17(7), 1979-1983 (2007-02-16)
A library of potential antifungal triazole-modified beta-methoxyacrylate analogues was designed and synthesized via a Cu(I)-catalyzed 1,3-dipolar alkyne-azide coupling reaction or 'click chemistry'. Subsequent biological screening revealed that some compounds displayed low to moderate antifungal activity toward pathogenic fungi and low

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service