Skip to Content
Merck
All Photos(1)

Documents

361844

Sigma-Aldrich

Chlorotris(triphenylphosphine)cobalt(I)

97%

Synonym(s):

CoCl(Ph3P)3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)3P]3CoCl
CAS Number:
Molecular Weight:
881.24
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

reaction suitability

core: cobalt
reagent type: catalyst

mp

135-139 °C (dec.) (lit.)

SMILES string

Cl[Co].c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9

InChI

1S/3C18H15P.ClH.Co/c3*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;/h3*1-15H;1H;/q;;;;+1/p-1

InChI key

RHNBOEDTKLKUEB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

A stoichiometric reducing agent employed in the radical dimerization of halogenated organic molecules.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sharanjeet K Bagal et al.
The Journal of organic chemistry, 69(26), 9100-9108 (2004-12-22)
The biomimetic synthesis of the bisesquiterpene lactones (+/-)-biatractylolide 1 and (+/-)-biepiasterolide 2 via dimerization of the captodative stabilized radical 8 is reported. Atractylon 7 has also been shown to be a possible intermediate during the biosynthesis of biatractylolide 1, biepiasterolide
Concise total synthesis of (-)-calycanthine, (+)-chimonanthine, and (+)-folicanthine.
Mohammad Movassaghi et al.
Angewandte Chemie (International ed. in English), 46(20), 3725-3728 (2007-04-07)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service