Skip to Content
Merck
All Photos(1)

Key Documents

416967

Sigma-Aldrich

Propargyl ether

98%

Synonym(s):

Dipropargyl ether, Dipropynyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HC≡CCH2)2O
CAS Number:
Molecular Weight:
94.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

liquid

refractive index

n20/D 1.442 (lit.)

bp

55-57 °C/65 mmHg (lit.)

density

0.914 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C#CCOCC#C

InChI

1S/C6H6O/c1-3-5-7-6-4-2/h1-2H,5-6H2

InChI key

HRDCVMSNCBAMAM-UHFFFAOYSA-N

General description

Propargylic ethers have been reported to participate in the preparation of various diindolylmethanes.[1]

Application

Propargyl ether was used for the preparation of polyferrocenes, via polyaddition with 1,1′-bis(azidoethyl)ferrocene.[2] It may be used for the preaparation of cyclic polystyrene.[3]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao Wang et al.
International journal of nanomedicine, 12, 4747-4762 (2017-07-26)
Vaccination is the most cost-effective means of infectious disease control. Although current influenza vaccines are effective in battling closely matched strains, such vaccines have major limitations such as the requirement to produce new vaccines every season, an egg-dependent production system
Cyclic polystyrene topologies via RAFT and CuAAC.
Hossain MD, et al.
Polym. Chem., 3(10), 2986-2995 (2012)
An Access Route to Polyferrocenes via Modular Conjugation.
Lang C, et al.
Macromolecular Chemistry and Physics, 212(8), 831-839 (2011)
Dongxu Shu et al.
Organic letters, 15(16), 4162-4165 (2013-08-06)
Various diindolylmethanes were prepared from propargylic ethers and substituted indoles via a platinum-catalyzed tandem indole annulation/arylation cascade. The resulting diindolylmethanes could be converted to natural product malassezin by formylation or indolo[3,2-b]carbazoles by cyclization.
Reggie L Hudson et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 233, 118206-118206 (2020-03-09)
The infrared (IR) spectrum of dipropargyl ether, (HC≡C-CH2)2O, has been reinvestigated for the compound's liquid, amorphous, and crystalline forms. The IR baseline changes and bandshape distortions seen in literature spectra have been considerably reduced by a different choice of conditions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service