Skip to Content
Merck
All Photos(1)

Key Documents

50062

Sigma-Aldrich

Glycidyl propargyl ether

technical, ≥90% (GC)

Synonym(s):

2,3-Epoxypropyl propargyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$259.00

$259.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
$259.00

About This Item

Empirical Formula (Hill Notation):
C6H8O2
CAS Number:
Molecular Weight:
112.13
Beilstein:
1098793
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$259.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical

Assay

≥90% (GC)

refractive index

n20/D 1.448

density

1.040 g/mL at 20 °C (lit.)

functional group

ether

SMILES string

C#CCOCC1CO1

InChI

1S/C6H8O2/c1-2-3-7-4-6-5-8-6/h1,6H,3-5H2

InChI key

SYFZCLMMUNCHNH-UHFFFAOYSA-N

Related Categories

General description

Glycidyl propargyl ether (GPE) is a glycidyl ether containing a terminal alkyne group.

Application

Glycidyl propargyl ether may be used in the synthesis of:
  • propargyl-functional poly(carbonate)s[1]
  • hyperbranched polyglycerol (hbPG)[2]
  • alkyne-functionalized polytetrahydrofuran (PTHF) diols[3]
  • dextran-based cyclodextrin polymers[4]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 1B - Resp. Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-step synthesis of multi-alkyne functional hyperbranched polyglycerols by copolymerization of glycidyl propargyl ether and glycidol.
Schull C, et al.
Polym. Chem., 4(17), 4730-4736 (2013)
Combining cationic ring-opening polymerization and click chemistry for the design of functionalized polyurethanes.
Basko M, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 49(7), 1597-1604 (2011)
Zoltán Fülöp et al.
Carbohydrate polymers, 97(2), 635-642 (2013-08-06)
Cyclodextrins and their water-soluble derivatives are excellent solubilizers and stabilizers and widely used as enabling pharmaceutical excipients. However, still new cyclodextrin derivatives are being designed and synthesized in an effort to improve their physicochemical properties. In this study physicochemical and
Propargyl-Functional Aliphatic Polycarbonate Obtained from Carbon Dioxide and Glycidyl Propargyl Ether.
Hilf J and Frey H.
Macromolecular Rapid Communications, 34(17), 1395-1400 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service