Skip to Content
Merck
All Photos(2)

Key Documents

415154

Sigma-Aldrich

Potassium tetracyanonickelate(II) hydrate

Synonym(s):

Dipotassium nickel(2+) tetracyanide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$280.00

$280.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$280.00

About This Item

Linear Formula:
K2Ni(CN)4 · xH2O
CAS Number:
Molecular Weight:
240.96 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

$280.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor pressure

740 mmHg ( 25 °C)(lit.)

Quality Level

form

powder

reaction suitability

reagent type: catalyst
core: nickel

density

1.875 g/mL (lit.)

SMILES string

[K+].[K+].[H]O[H].N#C[Ni--](C#N)(C#N)C#N

InChI

1S/4CN.2K.Ni.H2O/c4*1-2;;;;/h;;;;;;;1H2/q;;;;2*+1;-2;

InChI key

LORCEWUZCQZQEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassiumtetracyanonickelate (II) hydrate, also known as nickel(II) potassium cyanide, isa highly water-soluble compound used in electroplating and as a reagent inchemical synthesis.

Application


  • Electronic Structure: Study of the electronic structure of tetracyanonickelate(II) complexes, essential for applications in materials science and coordination chemistry (JJ Oppenheim, BJ McNicholas, J Miller, 2019).

Potassium tetracyanonickelate(II) hydrate can be used:
  • As a precursor for the synthesis of Hofmann-type metal-organic frameworks (MOFs) with high open metal sites density.
  • As a precursor in the synthesis of an anion-exchange resin for the removal of residual cyanide complexes in industrial plating wastewater.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service