Skip to Content
Merck
All Photos(2)

Key Documents

379166

Sigma-Aldrich

Potassium dicyanoargentate

Synonym(s):

Potassium silver(I) cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K[Ag(CN)2]
CAS Number:
Molecular Weight:
199.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder
Pricing and availability is not currently available.

grade

for analytical purposes

form

powder

reaction suitability

reagent type: catalyst
core: silver

SMILES string

[K+].N#C[Ag-]C#N

InChI

1S/2CN.Ag.K/c2*1-2;;/q;;-1;+1

InChI key

RQDBZJVQSLGBMI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dicyanoargentate ion is a versatile bridging ligand that has application for constructing multidimensional polymers.[1]

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soma, T. Iwamoto, T.
Inorganic Chemistry, 35, 1849-1849 (1996)
Fahad Ahmed Al-Abbasi et al.
Pakistan journal of pharmaceutical sciences, 31(4), 1431-1435 (2018-07-24)
The present study aimed at investigating the in-vitro oxidation of acrylonitrile (ACN) to cyanide (CN-) by prostaglandin H synthase (PHS). Detection of CN- is considered a marker for free radical intermediates involved in ACN-induced toxicity. First, most favorable circumstances for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service