Skip to Content
Merck
All Photos(2)

Key Documents

281557

Sigma-Aldrich

Potassium tetracyanoplatinate(II)

Synonym(s):

Dipotassium tetracyanoplatinate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$256.00

$256.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$256.00

About This Item

Linear Formula:
K2Pt(CN)4
CAS Number:
Molecular Weight:
377.34
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid

$256.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

form

solid

SMILES string

[K+].[K+].N#C[Pt--](C#N)(C#N)C#N

InChI

1S/4CN.2K.Pt/c4*1-2;;;/q;;;;2*+1;-2

InChI key

GPNDCURIWOPVTF-UHFFFAOYSA-N

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service