Skip to Content
Merck
All Photos(1)

Key Documents

902098

Sigma-Aldrich

4-Methacryloxyethyl trimellitic anhydride

Synonym(s):

(4-META)

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$231.00
25 G
$508.00

$231.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$231.00
25 G
$508.00

About This Item

Empirical Formula (Hill Notation):
C15H12O7
CAS Number:
Molecular Weight:
304.25
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$231.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

crystalline (solid )

color

white to off-white

storage temp.

2-8°C

SMILES string

O1C(=O)c2c(ccc(c2)C(=O)OCCOC(=O)C(=C)C)C1=O

InChI

1S/C15H12O7/c1-8(2)12(16)20-5-6-21-13(17)9-3-4-10-11(7-9)15(19)22-14(10)18/h3-4,7H,1,5-6H2,2H3

InChI key

RMCCONIRBZIDTH-UHFFFAOYSA-N

General description

4-Methacryloxyethyl trimellitic anhydride (4-META), a widely used dental adhesive, improves the bond strength between the metal and acrylic resin. It has an anhydride group that bonds with dental metal substrates and the methacrylate group that polymerizes and copolymerizes with monomeric composite resin.[1][2]

Application

4-META can be used in the formation of resin to fix the vertical root fracture by root fragment bonding.[3]
4-Methacryloxyethyl trimellitic anhydride (4-META) is a methacylate monomer most commonly used as an adhesion promoter in dental resins. 4-META contains both hydrophilic and hydrophobic groups, promoting the diffusion of other dental resin monomers into the tooth structure to form a hybrid, resin-impregnated layer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shear bond strength between autopolymerizing acrylic resin and Co-Cr alloy using different primers
Sanohkan S, et al.
Dental Materials Journal, 31(5) (2012)
Successful treatment of vertical root fracture through intentional replantation and root fragment bonding with 4-META/MMA-TBB resin
Okaguchi M, et al.
Journal of the Formosan Medical Association = Taiwan Yi Zhi, 118(3), 671-678 (2019)
Evaluation of biological, physical and chemical properties of mineral trioxide aggregate mixed with 4-META/MMA-TBB
Kaul R, et al.
Indian Journal of Dental Research, 24(4), 418-418 (2013)
Jeffrey Chai Chang et al.
The Journal of prosthetic dentistry, 87(2), 216-224 (2002-02-21)
4-META (4-methacryloyloxyethy trimellitate anhydride) was invented and patented in Japan in January 1979 and patented in the United States in April 1979. For the first 8 years, 4-META dental products (manufactured by Sun Medical Co, Kyoto, Japan) were not readily

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service