Skip to Content
Merck
All Photos(1)

Key Documents

795739

Sigma-Aldrich

11-Phosphonoundecyl acrylate

Synonym(s):

(11-(Acryloyloxy)undecyl)phosphonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$204.00

$204.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$204.00

About This Item

Empirical Formula (Hill Notation):
C14H27O5P
CAS Number:
Molecular Weight:
306.33
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$204.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

SMILES string

O=C(C=C)OCCCCCCCCCCCP(O)(O)=O

InChI

1S/C14H27O5P/c1-2-14(15)19-12-10-8-6-4-3-5-7-9-11-13-20(16,17)18/h2H,1,3-13H2,(H2,16,17,18)

InChI key

RLAKYYIYWIWCED-UHFFFAOYSA-N

General description

11-Phosphonoundecyl acrylate is a bifuncational ligand that forms a self-assembled monolayer (SAM) on a variety of substrates. It binds with the surface metal atoms by attaching the phosphonic acid head groups. [1]

Application

Tuning of surface energy, wetting, and work function of metal oxides. Allows further chemistry on surface.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mechanoresponsive Polymerized Liquid Metal Networks
Thrasher C, et al.
Advanced Materials, 1903864-1903864 (2019)
Carl J Thrasher et al.
Advanced materials (Deerfield Beach, Fla.), 31(40), e1903864-e1903864 (2019-08-14)
Room-temperature liquid metals, such as nontoxic gallium alloys, show enormous promise to revolutionize stretchable electronics for next-generation soft robotic, e-skin, and wearable technologies. Core-shell particles of liquid metal with surface-bound acrylate ligands are synthesized and polymerized together to create cross-linked

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service