Skip to Content
Merck
All Photos(1)

Key Documents

405825

Sigma-Aldrich

Methyl 3,5-dihydroxyphenylacetate

97%

Synonym(s):

(3,5-Dihydroxyphenyl)acetic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$114.00
5 G
$338.00

$114.00


Estimated to ship onApril 14, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$114.00
5 G
$338.00

About This Item

Linear Formula:
(HO)2C6H3CH2CO2CH3
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$114.00


Estimated to ship onApril 14, 2025


Request a Bulk Order

Quality Level

Assay

97%

mp

109-112 °C (lit.)

functional group

ester

SMILES string

COC(=O)Cc1cc(O)cc(O)c1

InChI

1S/C9H10O4/c1-13-9(12)4-6-2-7(10)5-8(11)3-6/h2-3,5,10-11H,4H2,1H3

InChI key

LMLSBPHXMGSGCR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl 3,5-dihydroxyphenylacetate is a phenylacetic acid derivative. It is a known metabolite of Curvularia Siddiqui. It has been extracted from the culture mycelia of Curvularia lunata supported on culture medium (consisting of yeast, malt extract and glucose).[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gouri B Varma et al.
Phytochemistry, 67(17), 1925-1930 (2006-08-04)
Phenylacetic acid derivatives, methyl 2-acetyl-3,5-dihydroxyphenylacetate (1) and methyl 2-acetyl-5-hydroxy-3-methoxyphenylacetate (3); curvulin or ethyl 2-acetyl-3,5-dihydroxyphenylacetate (4), a known metabolite of Curvularia siddiqui, and 4-epiradicinol (5) have been isolated from the culture mycelia of Curvularia lunata grown on YMG, a medium consisting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service