Skip to Content
Merck
All Photos(2)

Key Documents

M11103

Sigma-Aldrich

4-Methoxybenzylamine

98%

Synonym(s):

(4-Methoxyphenyl)methanamine, (p-Methoxyphenyl)methanamine, 1-Aminomethyl-4-methoxybenzene, 4-Methoxybenzenemethanamine, Anisylamine, p-Methoxybenzylamine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$191.00
100 G
$716.00

$191.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$191.00
100 G
$716.00

About This Item

Linear Formula:
CH3OC6H4CH2NH2
CAS Number:
Molecular Weight:
137.18
Beilstein:
508206
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$191.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.546 (lit.)

bp

236-237 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(CN)cc1

InChI

1S/C8H11NO/c1-10-8-4-2-7(6-9)3-5-8/h2-5H,6,9H2,1H3

InChI key

IDPURXSQCKYKIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Methoxybenzylamine is a primary amine and can be used for:
  • Amination reaction of functionalized aryl bromides.[1]
  • Synthesis of functionalized organopolyphosphazenes for in vivo applications.[2]
  • Synthesis of analogs of myoseverin, 8-azapurine, for their antiproliferative activities.[3]
  • Studying the deprotection procedures of amino protecting groups such as 2-nitro- and 2,4-dinitrobenzenesulfonamides.[4]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Jonsson et al.
Biochemistry, 33(49), 14871-14878 (1994-12-13)
Competitive kH/kT and kD/kT kinetic isotope effects on p-methoxybenzylamine oxidation by the 8 alpha-S-cysteinyl flavin adenine dinucleotide (FAD)-dependent enzyme monoamine oxidase B (MAO-B) have been measured as a function of temperature. At pH 7.5, exponents relating observed kH/kT and kD/kT
2-Nitro-and 2, 4-dinitrobenzenesulfonamides as protecting groups for primary amines.
Nihei KI, et al.
Synlett, 2001(07), 1167-1169 (2001)
Synthesis and biological activity of 8-azapurine and pyrazolo [4, 3-d] pyrimidine analogues of myoseverin.
Krystof V, et al.
European Journal of Medicinal Chemistry, 41(12), 1405-1411 (2006)
Anna Berlicka et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 9(10), 1359-1366 (2010-09-08)
A variety of primary and secondary benzylic amines were oxidized efficiently to N-benzylidenebenzylamines and imines, respectively, using 2,7,12,17-tetrapropylporphycene (H(2)TPrPc) photocatalyst and blue light emitting diodes (LEDs). The photooxidation of 4-methoxybenzylamine in the presence of H(2)TPrPc and its tin(IV) complex Sn(TPrPc)Cl(2)
Synthesis and characterization of novel polyorganophosphazenes substituted with 4-methoxybenzylamine and 4-methoxyphenethylamine for in vitro release of indomethacin and 5-fluorouracil.
Gudasi KB, et al.
Reactive and Functional Polymers, 66(10), 1149-1157 (2006)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service